Methyl (2S,4S)-4-[3,5-bis(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate

Supplier: Matrix Scientific

023614-500MG
101816-348EA 235.75 USD
101816-348
Methyl (2S,4S)-4-[3,5-bis(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate
Methyl (2S,4S)-4-[3,5-bis(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₃F₆NO₃
MDL Number: MFCD08688184
CAS Number: 1135225-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR