(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023612-500MG
101816-344EA 156.4 USD
101816-344
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-fluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉ClFNO₅
MDL Number: MFCD08687080
CAS Number: 1135226-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR