(2S,4S)-4-(4-Bromo-2-methylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023611-500MG
101816-342EA 235.75 USD
101816-342
(2S,4S)-4-(4-Bromo-2-methylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Bromo-2-methylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂BrNO₅
MDL Number: MFCD08687024
CAS Number: 1354487-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR