(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023609-500MG
101816-338EA 315.38 USD
101816-338
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O₇
MDL Number: MFCD08687085
CAS Number: 1217655-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR