(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-difluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023606-500MG
101816-332EA 315.38 USD
101816-332
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-difluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-difluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉F₂NO₅
MDL Number: MFCD08687095
CAS Number: 869682-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR