(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-ethyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023604-500MG
101816-330EA 315.38 USD
101816-330
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-ethyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-ethyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₇NO₆
MDL Number: MFCD08687107
CAS Number: 869682-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR