(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(2-methoxyethyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023599-500MG
101816-320EA 235.75 USD
101816-320
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(2-methoxyethyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(2-methoxyethyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₇NO₆
MDL Number: MFCD08687122
CAS Number: 1217603-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR