(2S,4S)-4-(4-Bromo-2-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023597-500MG
101816-316EA 402.5 USD
101816-316
(2S,4S)-4-(4-Bromo-2-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Bromo-2-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉BrFNO₅
MDL Number: MFCD08686998
CAS Number: 1135227-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR