(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023595-500MG
101816-312EA 315.38 USD
101816-312
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropyl-3-methylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₉NO₅
MDL Number: MFCD08687117
CAS Number: 1354487-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR