(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
023593-500MG
101816-308EA
315.38
USD
101816-308
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-difluorophenoxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₉F₂NO₅ |
MDL Number:
MFCD08687094 CAS Number: 317357-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...