(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023591-500MG
101816-304EA 315.38 USD
101816-304
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₇
MDL Number: MFCD08687135
CAS Number: 218943-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR