(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023590-500MG
101816-302EA 315.38 USD
101816-302
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-nitrophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₇
MDL Number: MFCD08687134
CAS Number: 1354484-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR