(2S,4S)-1-(tert-Butoxycarbonyl)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
023589-500MG
101816-300EA
538.46
USD
101816-300
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(8-quinolinyloxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₉H₂₂N₂O₅ |
MDL Number:
MFCD08687141 CAS Number: 1135208-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...