(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-pyridinyloxy)-2-pyrrolidine carboxylic acid

Supplier: Matrix Scientific

023588-500MG
101816-298EA 402.5 USD
101816-298
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-pyridinyloxy)-2-pyrrolidine carboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-pyridinyloxy)-2-pyrrolidine carboxylic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₅
MDL Number: MFCD08687140
CAS Number: 1135195-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR