(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-naphthyloxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023586-500MG
101816-294EA 315.38 USD
101816-294
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-naphthyloxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-naphthyloxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₃NO₅
MDL Number: MFCD06656449
CAS Number: 317357-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR