(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,3-dihydro-1H-inden-5-yloxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023583-500MG
101816-288EA 235.75 USD
101816-288
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,3-dihydro-1H-inden-5-yloxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,3-dihydro-1H-inden-5-yloxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₅NO₅
MDL Number: MFCD08687097
CAS Number: 1354486-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR