(2S,4S)-4-(4-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023581-500MG
101816-284EA 315.38 USD
101816-284
(2S,4S)-4-(4-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₇NO₅
MDL Number: MFCD08686887
CAS Number: 1217819-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR