(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methoxy-4-methylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023580-500MG
101816-282EA 235.75 USD
101816-282
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methoxy-4-methylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methoxy-4-methylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₅NO₆
MDL Number: MFCD08687123
CAS Number: 1354486-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR