(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(sec-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023578-500MG
101816-278EA 247.25 USD
101816-278
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(sec-butyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(sec-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₉NO₅
MDL Number: MFCD08687071
CAS Number: 1217545-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR