(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023577-500MG
101816-276EA 315.38 USD
101816-276
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₇NO₅
MDL Number: MFCD08687121
CAS Number: 1354486-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR