(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(tert-pentyl)phenoxy]-2-pyrrolidine carboxylic acid

Supplier: Matrix Scientific

023576-500MG
101816-274EA 538.46 USD
101816-274
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(tert-pentyl)phenoxy]-2-pyrrolidine carboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(tert-pentyl)phenoxy]-2-pyrrolidine carboxylic acid

Hazard: Irritant

Formula: C₂₁H₃₁NO₅
MDL Number: MFCD08687136
CAS Number: 1217725-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR