(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
023572-500MG
101816-266EA
315.38
USD
101816-266
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₇H₂₀F₃NO₅ |
MDL Number:
MFCD08687146 CAS Number: 1135226-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...