(2S,4S)-4-[4-(Benzyloxy)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023568-500MG
101816-258EA 315.38 USD
101816-258
(2S,4S)-4-[4-(Benzyloxy)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-[4-(Benzyloxy)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₇NO₆
MDL Number: MFCD08686874
CAS Number: 1354487-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR