(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-phenoxyphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023566-500MG
101816-254EA 495.38 USD
101816-254
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-phenoxyphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-phenoxyphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₆
MDL Number: MFCD08687137
CAS Number: 1217845-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR