(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023563-500MG
101816-248EA 402.5 USD
101816-248
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂ClNO₅
MDL Number: MFCD08687083
CAS Number: 1217723-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR