(2S,4S)-4-(2-Bromo-4-chlorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023562-500MG
101816-246EA 235.75 USD
101816-246
(2S,4S)-4-(2-Bromo-4-chlorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Bromo-4-chlorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉BrClNO₅
MDL Number: MFCD08686969
CAS Number: 1354484-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR