(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chlorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023561-500MG
101816-244EA 461.54 USD
101816-244
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chlorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chlorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀ClNO₅
MDL Number: MFCD08687088
CAS Number: 686766-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR