(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023560-500MG
101816-242EA 538.46 USD
101816-242
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀FNO₅
MDL Number: MFCD08687114
CAS Number: 869681-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR