(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023556-500MG
101816-234EA 315.38 USD
101816-234
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₅NO₅
MDL Number: MFCD08687102
CAS Number: 1354486-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR