(2S,4S)-1-(tert-Butoxycarbonyl)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023553-500MG
101816-228EA 235.75 USD
101816-228
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₉NO₅
MDL Number: MFCD08687072
CAS Number: 1217838-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR