(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023552-500MG
101816-226EA 235.75 USD
101816-226
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-methoxyphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₃NO₆
MDL Number: MFCD08687125
CAS Number: 1354486-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR