(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dichlorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023548-500MG
101816-218EA 538.46 USD
101816-218
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dichlorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,4-dichlorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉Cl₂NO₅
MDL Number: MFCD08687093
CAS Number: 1217803-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR