(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023545-500MG
101816-212EA 315.38 USD
101816-212
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀FNO₅
MDL Number: MFCD08687113
CAS Number: 869681-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR