(2S,4S)-4-(2-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023540-500MG
101816-202EA 538.46 USD
101816-202
(2S,4S)-4-(2-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Benzylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₇NO₅
MDL Number: MFCD08686886
CAS Number: 1217636-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR