(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023534-500MG
101816-190EA 235.75 USD
101816-190
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₃NO₅
MDL Number: MFCD08687129
CAS Number: 869681-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR