(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023530-500MG
101816-182EA 315.38 USD
101816-182
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-ethoxyphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₅NO₆
MDL Number: MFCD08687104
CAS Number: 869682-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR