(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023527-500MG
101816-176EA 315.38 USD
101816-176
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,5-dichlorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉Cl₂NO₅
MDL Number: MFCD08687092
CAS Number: 1354486-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR