(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023525-500MG
101816-172EA 235.75 USD
101816-172
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉ClFNO₅
MDL Number: MFCD08687077
CAS Number: 1354486-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR