(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chlorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023522-500MG
101816-166EA 315.38 USD
101816-166
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chlorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chlorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀ClNO₅
MDL Number: MFCD08687086
CAS Number: 869682-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR