2-[(4-Methylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023517-500MG
101816-156EA 402.5 USD
101816-156
2-[(4-Methylphenoxy)methyl]piperidine
2-[(4-Methylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD06248325
CAS Number: 883547-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR