2-[(4-Methoxyphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023515-500MG
101816-154EA 315.38 USD
101816-154
2-[(4-Methoxyphenoxy)methyl]piperidine
2-[(4-Methoxyphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD02663689
CAS Number: 383128-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR