2-[(3-Methoxyphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023514-500MG
101816-152EA 453.85 USD
101816-152
2-[(3-Methoxyphenoxy)methyl]piperidine
2-[(3-Methoxyphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD08688082
CAS Number: 946727-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR