3-[(1-Naphthyloxy)methyl]pyrrolidine

Supplier: Matrix Scientific

023510-500MG
101816-146EA 315.38 USD
101816-146
3-[(1-Naphthyloxy)methyl]pyrrolidine
3-[(1-Naphthyloxy)methyl]pyrrolidine

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD08688431
CAS Number: 858934-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR