3-[(4-Methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023508-500MG
101816-142EA 538.46 USD
101816-142
3-[(4-Methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD12405063
CAS Number: 1185298-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR