4-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

023507-500MG
101816-140EA 538.46 USD
101816-140
4-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride
4-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD09997934
CAS Number: 1185298-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR