3-[(4-Methoxyphenoxy)methyl]pyrrolidine

Supplier: Matrix Scientific

023506-500MG
101816-138EA 235.75 USD
101816-138
3-[(4-Methoxyphenoxy)methyl]pyrrolidine
3-[(4-Methoxyphenoxy)methyl]pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD08688086
CAS Number: 933736-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR