3-[(3-Methoxyphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023503-500MG
101816-132EA 538.46 USD
101816-132
3-[(3-Methoxyphenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Methoxyphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560449
CAS Number: 1220037-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR