3-[(3-Fluorophenoxy)methyl]pyrrolidine

Supplier: Matrix Scientific

023502-500MG
101816-130EA 402.5 USD
101816-130
3-[(3-Fluorophenoxy)methyl]pyrrolidine
3-[(3-Fluorophenoxy)methyl]pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₄FNO
MDL Number: MFCD08687898
CAS Number: 946760-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR