3-(Phenoxymethyl)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023501-500MG
101816-128EA 315.38 USD
101816-128
3-(Phenoxymethyl)pyrrolidine hydrochloride
3-(Phenoxymethyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD09997929
CAS Number: 1185299-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR