3-[(Benzyloxy)methyl]pyrrolidine

Supplier: Matrix Scientific

023499-500MG
101816-126EA 402.5 USD
101816-126
3-[(Benzyloxy)methyl]pyrrolidine
3-[(Benzyloxy)methyl]pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD08686873
CAS Number: 933758-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR